d-Desthiobiotin 97%

Supplier: Ambeed
Warning

Synonyms: 5-Methyl-2-oxo-4-imidazolidinehexanoic acid, 6-((4R,5S)-5-Methyl-2-oxoimidazolidin-4-yl)hexanoic acid, Desthiobiotin

A558564-100MG A558564-1G A558564-250MG A558564-5G A558564-10G
76696-198EA 98.67 USD
76696-198 76696-200 76696-202 77551-678 77602-374
d-Desthiobiotin 97%
d-Desthiobiotin

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₃
MW: 214.26 g/mol
MDL Number: MFCD00213804
CAS Number: 533-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR