Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl ethoxymethylene-3-oxo-4,4,4-trifluorobutyrate, 3-Ethoxy-2-(2,2,2-trifluoroacetyl)acrylic Acid Ethyl Ester, Ethyl 3-Ethoxy-2-(2,2,2-trifluoroacetyl)acrylate, 2-(Ethoxymethylene)-4,4,4-trifluoro-3-oxobutyric Acid Ethyl Ester

A558088-100G A558088-10G A558088-1G A558088-25G A558088-500G A558088-5G
76696-186EA 187 USD
76696-186 76696-188 76696-190 76696-192 76696-194 76696-196
Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate 97%
Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate

Precaución: for Research Use Only

Formula: C₉H₁₁F₃O₄
MDL Number: MFCD02677683
CAS Number: 571-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR