Bromotriphenylethylene 98%

Supplier: Ambeed
Warning

A555106-100G A555106-25G A555106-5G
76696-166EA 108 USD
76696-166 76696-168 76696-170
Bromotriphenylethylene 98%
Bromotriphenylethylene

Precaución: for Research Use Only

Formula: (C₆H₅)₂CCBrC₆H₅
MDL Number: MFCD00000135
CAS Number: 1607-57-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR