(3-Methoxyphenyl)acetonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methoxybenzyl cyanide, 2-(3-Methoxyphenyl)acetonitrile

A552650-100G A552650-10G A552650-25G A552650-500G
76696-158EA 48 USD
76696-158 76696-160 76696-162 76696-164
(3-Methoxyphenyl)acetonitrile 98%
(3-Methoxyphenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₉H₉NO
MDL Number: MFCD00001910
CAS Number: 19924-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR