11-(1-Methylpiperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine dimaleate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Azatadine dimaleate
A550669-10MG
A550669-250MG
A550669-50MG
A550669-1MG
76696-124EA
6
USD
76696-124
76696-126
76696-128
77602-328
11-(1-Methylpiperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine dimaleate 98%
11-(1-Methylpiperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine dimaleate
Precaución: for Research Use Only
Formula:
C₂₈H₃₀N₂O₈ MW: 522.54 g/mol |
MDL Number:
MFCD00242593 CAS Number: 3978-86-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...