Methyl-2-amino-3-chlorobenzoate 97%

Supplier: Ambeed
Warning

A548514-100G A548514-10G A548514-25G A548514-5G
76696-106EA 33 USD
76696-106 76696-108 76696-110 76696-112
Methyl-2-amino-3-chlorobenzoate 97%
Methyl 2-amino-3-chlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MDL Number: MFCD06797833
CAS Number: 77820-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR