4-Methoxybenzyl acetate 98%

Supplier: Ambeed
Warning

Synonyms: Acetic Acid 4-Methoxybenzyl Ester, Anisyl Acetate, Acetic Acid Anisyl Ester

A548074-100G A548074-500G
76696-102EA 17 USD
76696-102 76696-104
4-Methoxybenzyl acetate 98%
4-Methoxybenzyl acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.20 g/mol
MDL Number: MFCD00038509
CAS Number: 104-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR