3-(2-Benzimidazolyl)-7-(diethylamino)coumarin 98%

Supplier: Ambeed
Warning

Synonyms: Coumarin 7, 3-(1H-Benzo[d]imidazol-2-yl)-7-(diethylamino)-2H-chromen-2-one

A544307-1G A544307-250MG A544307-5G
76696-076EA 75 USD
76696-076 76696-078 77572-728
3-(2-Benzimidazolyl)-7-(diethylamino)coumarin 98%
3-(2-Benzimidazolyl)-7-(diethylamino)coumarin

Precaución: for Research Use Only

Formula: C₂₀H₁₉N₃O₂
MW: 333.3838 g/mol
MDL Number: MFCD00051348
CAS Number: 27425-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR