Cyclohexanesulfonyl chloride 95%

Supplier: Ambeed
Danger

A543354-1G A543354-5G
76696-060EA 28 USD
76696-060 77572-724
Cyclohexanesulfonyl chloride 95%
Cyclohexanesulfonyl chloride

Precaución: for Research Use Only

Formula: C₆H₁₁ClO₂S
MDL Number: MFCD04117775
CAS Number: 4837-38-1
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR