Cysteaminium chloride 98%

Supplier: Ambeed
Warning

A542078-100G A542078-25G A542078-5G A542078-500G
76696-054EA 23 USD
76696-054 76696-056 76696-058 77551-644
Cysteaminium chloride 98%
Cysteaminium chloride
Formula: C₂H₇NS·HCl
MDL Number: MFCD00012904
CAS Number: 156-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR