prim-O-Glucosylcimifugin 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(2-Hydroxypropan-2-yl)-4-methoxy-7-((((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)-2H-furo[3,2-g]chromen-5(3H)-one

A540157-100MG A540157-1G A540157-250MG
76696-048EA 107.16 USD
76696-048 76696-050 76696-052
prim-O-Glucosylcimifugin 98%
prim-O-Glucosylcimifugin

Precaución: for Research Use Only

Formula: C₂₂H₂₈O₁₁
MW: 468.46 g/mol
MDL Number: MFCD09037394
CAS Number: 80681-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR