Veratrohydrazide 98%

Supplier: Ambeed
Warning

A540023-1G A540023-25G A540023-5G
76696-036EA 11 USD
76696-036 76696-038 76696-040
Veratrohydrazide 98%
Veratrohydrazide

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₃
MDL Number: MFCD00017061
CAS Number: 41764-74-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR