8-Phenyl-1-octanol 97%

Supplier: Ambeed
Warning

Synonyms: 8-Phenyloctan-1-ol

A539018-1G A539018-250MG A539018-5G
76696-012EA 78.51 USD
76696-012 76696-014 76696-016
8-Phenyl-1-octanol 97%
8-Phenyl-1-octanol

Precaución: for Research Use Only

Formula: C₁₄H₂₂O
MW: 206.3239 g/mol
MDL Number: MFCD00040918
CAS Number: 10472-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR