(S)-Benzyl 2-(pyrrolidine-1-carbonyl)pyrrolidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A538957-1G A538957-5G
76696-008EA 68.96 USD
76696-008 76696-010
(S)-Benzyl 2-(pyrrolidine-1-carbonyl)pyrrolidine-1-carboxylate 97%
(S)-Benzyl 2-(pyrrolidine-1-carbonyl)pyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₂N₂O₃
MW: 302.36 g/mol
MDL Number: MFCD00799477
CAS Number: 50888-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR