Behenic acid 85%
Supplier: Ambeed
|
Warning
|
Synonyms:
Docosanoic acid
A538500-1000G
A538500-100G
A538500-10G
A538500-500G
A538500-25G
76696-000EA
51
USD
76696-000
76696-002
76696-004
76696-006
77307-764
Behenic acid 85%
Behenic acid
Precaución: for Research Use Only
Formula:
C₂₂H₄₄O₂ MW: 340.59 g/mol Boiling Pt: 306 Melting Pt: 80 |
MDL Number:
MFCD00002807 CAS Number: 112-85-6 Merck Index: 13,01024 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...