Cbz-1-amino-1-cyclopentanecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1-(Cbz-amino)cyclopentanecarboxylic Acid

A537727-100G A537727-1G A537727-25G A537727-5G
76695-978EA 776.58 USD
76695-978 76695-980 76695-982 76695-984
Cbz-1-amino-1-cyclopentanecarboxylic acid 98%
Cbz-1-amino-1-cyclopentanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₄
MW: 263.293 g/mol
MDL Number: MFCD02094399
CAS Number: 17191-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR