D-erythro-Sphingosine 98%

Supplier: Ambeed
Warning

A536742-1MG A536742-1G A536742-250MG A536742-100MG A536742-5G
76695-976EA 7.43 USD
76695-976 77569-038 77569-040 77572-700 77627-090
D-erythro-Sphingosine 98%
D-erythro-Sphingosine

Precaución: For research use only.

Formula: C₁₈H₃₇NO₂
MDL Number: MFCD00036751
CAS Number: 123-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR