Bis(4-methoxyphenyl)phenylphosphine 98%

Supplier: Ambeed
Warning

Synonyms: Bis(4-methoxyphenyl)(phenyl)phosphine

A532415-1G A532415-25G A532415-5G
76695-964EA 21.22 USD
76695-964 76695-966 76695-968
Bis(4-methoxyphenyl)phenylphosphine 98%
Bis(4-methoxyphenyl)phenylphosphine

Precaución: for Research Use Only

Formula: C₂₀H₁₉O₂P
MW: 322.34 g/mol
MDL Number: MFCD00048993
CAS Number: 14180-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR