Ammonium dihydrogen phosphate 99%

Supplier: Ambeed
Warning

Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

A532100-1000G A532100-500G
76695-958EA 16.98 USD
76695-958 76695-960 76695-962
Ammonium dihydrogen phosphate 99%
Ammonium dihydrogen phosphate

Precaución: for Research Use Only

Formula: NH₄H₂PO₄
MW: 115.03 g/mol
MDL Number: MFCD00003396
CAS Number: 7722-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR