Benzyloxyacetaldehyde diethyl acetal 98%

Supplier: Ambeed
Warning

A530626-100G A530626-1G A530626-5G
76695-946EA 265 USD
76695-946 76695-948 76695-950
Benzyloxyacetaldehyde diethyl acetal 98%
Benzyloxyacetaldehyde diethyl acetal

Precaución: for Research Use Only

Formula: C₁₃H₂₀O₃
MDL Number: MFCD00051786
CAS Number: 42783-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR