Clonidine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 2-(2,6-Dichlorophenylamino)-2-imidazoline Hydrochloride, N-(2,6-Dichlorophenyl)-4,5-dihydro-1H-imidazol-2-amine hydrochloride, 2-(2,6-Dichloroanilino)-2-imidazoline Hydrochloride

A530479-100MG A530479-1G A530479-250MG A530479-25G A530479-25MG A530479-5G
76695-934EA 8 USD
76695-934 76695-936 76695-938 76695-940 76695-942 76695-944
Clonidine hydrochloride 98%
Clonidine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₉Cl₂N₃
MW: 266.55 g/mol
MDL Number: MFCD00036705
CAS Number: 4205-91-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR