(R)-2-Acetamido-3-(acetylthio)propanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: Dacisteine

A530388-1G A530388-250MG A530388-100MG A530388-5G
76695-930EA 95 USD
76695-930 76695-932 77602-258 77627-080
(R)-2-Acetamido-3-(acetylthio)propanoic acid 97%
(R)-2-Acetamido-3-(acetylthio)propanoic acid

Precaución: for Research Use Only

Formula: C₇H₁₁NO₄S
MW: 205.23 g/mol
MDL Number: MFCD00152052
CAS Number: 18725-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR