9-Hydroxy-4-methoxyacridine 95%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxyacridin-9-ol

A529764-100MG A529764-1G A529764-5G
76695-926EA 6 USD
76695-926 76695-928 77878-316
9-Hydroxy-4-methoxyacridine 95%
9-Hydroxy-4-methoxyacridine

Precaución: for Research Use Only

Formula: C₁₄H₁₁NO₂
MW: 225.25 g/mol
MDL Number: MFCD00005027
CAS Number: 35308-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR