6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A529081-1G A529081-5G
76695-922EA 9.55 USD
76695-922 76695-924
6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid 98%
6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₆N₂O₃
MW: 142.12 g/mol
MDL Number: MFCD00052030
CAS Number: 27372-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR