Bis(2-methoxyethyl)amine 98%

Supplier: Ambeed
Danger

A528779-250MG A528779-25G A528779-1G A528779-5G
76695-918EA 5 USD
76695-918 76695-920 77627-076 77627-078
Bis(2-methoxyethyl)amine 98%
Bis(2-methoxyethyl)amine

Precaución: for Research Use Only

Formula: C₆H₁₅NO₂
MDL Number: MFCD00025906
CAS Number: 111-95-5
UN: 2734
ADR: 8,3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR