Allyloxytrimethylsilane 98%

Supplier: Ambeed
Danger

A527234-25G A527234-5G
76695-908EA 37.14 USD
76695-908 76695-910
Allyloxytrimethylsilane 98%
Allyloxytrimethylsilane

Precaución: for Research Use Only

Formula: C₆H₁₄OSi
MW: 130.26 g/mol
MDL Number: MFCD00008643
CAS Number: 18146-00-4
UN: 2924
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR