D-(+)-Turanose 98%

Supplier: Ambeed
Warning

Synonyms: (3S,4R,5R)-1,4,5,6-Tetrahydroxy-3-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexan-2-one, Turanose

A526766-1G A526766-250MG A526766-5G
76695-880EA 58.35 USD
76695-880 76695-882 76695-884
D-(+)-Turanose 98%
D(+)-Turanose

Precaución: for Research Use Only

Formula: C₁₂H₂₂O₁₁
MW: 342.30 g/mol
MDL Number: MFCD00006606
CAS Number: 547-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR