Ethyl 4-(4-formyl-3-hydroxyphenoxy)butanoate 97%

Supplier: Ambeed
Warning

A524243-1G A524243-5G
76695-868EA 18.04 USD
76695-868 76695-870
Ethyl 4-(4-formyl-3-hydroxyphenoxy)butanoate 97%
Ethyl 4-(4-formyl-3-hydroxyphenoxy)butanoate

Precaución: for Research Use Only

Formula: C₁₃H₁₆O₅
MW: 252.26 g/mol
MDL Number: MFCD29765746
CAS Number: 152942-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR