Citronellyl acetate 95%

Supplier: Ambeed
Danger

Synonyms: 3,7-Dimethyloct-6-en-1-yl acetate, 3,7-Dimethyl-6-octen-1-ol Acetate

A522404-25G A522404-100G A522404-500G
76695-860EA 11 USD
76695-860 77602-234 77602-236
Citronellyl acetate 95%
Citronellyl acetate

Precaución: for Research Use Only

Formula: C₁₂H₂₂O₂
MDL Number: MFCD00015039
CAS Number: 150-84-5
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR