Alloxan monohydrate 98% (by titrimetric analysis)

Supplier: Ambeed
Warning

Synonyms: Pyrimidine-2,4,5,6(1H,3H)-tetraone hydrate

A521632-25G A521632-5G A521632-100G A521632-1G
76695-844EA 61 USD
76695-844 76695-846 77627-056 77627-058
Alloxan monohydrate 98% (by titrimetric analysis)
Alloxan monohydrate

Precaución: for Research Use Only

Formula: C₄H₄N₂O₅
MW: 160.09 g/mol
MDL Number: MFCD00006031
CAS Number: 2244-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR