9,10-Bis(2-(4,5-dihydro-1H-imidazol-2-yl)hydrazono)methyl)anthracene 95+%

Supplier: Ambeed
Warning

Synonyms: Bisantrene

A518842-10MG A518842-1MG A518842-25MG A518842-50MG
76695-824EA 125.19 USD
76695-824 76695-826 76695-828 76695-830
9,10-Bis(2-(4,5-dihydro-1H-imidazol-2-yl)hydrazono)methyl)anthracene 95+%
9,10-Bis(2-(4,5-dihydro-1H-imidazol-2-yl)hydrazono)methyl)anthracene

Precaución: for Research Use Only

Formula: C₂₂H₂₂N₈
MW: 398.46 g/mol
MDL Number: MFCD00872137
CAS Number: 78186-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR