Creatine (N-amidinosarcosine) 95%

Supplier: Ambeed
Warning

Synonyms: 2-(1-Methylguanidino)acetic acid, N-Amidinosarcosine, Creatine

A518760-100G A518760-25G A518760-500G
76695-818EA 13.8 USD
76695-818 76695-820 76695-822
Creatine (N-amidinosarcosine) 95%
Creatine (N-amidinosarcosine)

Precaución: for Research Use Only

Formula: C₄H₉N₃O₂
MW: 131.13 g/mol
MDL Number: MFCD00004282
CAS Number: 57-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR