Di(1-adamantyl)-n-butylphosphine 97%

Supplier: Ambeed
Danger

A516771-1G A516771-250MG A516771-5G A516771-10G A516771-25G A516771-100G
76695-812EA 8 USD
76695-812 76695-814 76695-816 77551-546 77551-548 77627-038
Di(1-adamantyl)-n-butylphosphine 97%
Di(1-adamantyl)-n-butylphosphine

Precaución: for Research Use Only

Formula: C₂₄H₃₉P
MDL Number: MFCD05861606
CAS Number: 321921-71-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR