Ethyl oxo(2-oxocyclopent-1-yl)acetate 96%

Supplier: Ambeed
Warning

A511545-1G A511545-250MG A511545-5G
76695-786EA 53 USD
76695-786 76695-788 76695-790
Ethyl oxo(2-oxocyclopent-1-yl)acetate 96%
Ethyl oxo(2-oxocyclopent-1-yl)acetate

Precaución: for Research Use Only

Formula: C₉H₁₂O₄
MDL Number: MFCD00019311
CAS Number: 39163-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR