2,4-Dimethoxy-3-methylbenzaldehyde 97%

Supplier: Ambeed
Warning

A510975-1G A510975-250MG A510975-5G
76695-774EA 13 USD
76695-774 76695-776 76695-778
2,4-Dimethoxy-3-methylbenzaldehyde 97%
2,4-Dimethoxy-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MDL Number: MFCD00011489
CAS Number: 7149-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR