7-Hydroxyquinolin-2(1H)-one 95%

Supplier: Ambeed
Warning

Synonyms: 2,7-Dihydroxyquinoline, 7-Hydroxyquinolin-2(1H)-one

A510778-10G A510778-1G A510778-25G A510778-5G
76695-766EA 28.65 USD
76695-766 76695-768 76695-770 76695-772
7-Hydroxyquinolin-2(1H)-one 95%
7-Hydroxyquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MW: 161.157 g/mol
MDL Number: MFCD04117979
CAS Number: 70500-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR