Copper(II) acetate monohydrate 97%

Supplier: Ambeed
Danger

Synonyms: Copper diacetate monohydrate, Cupric acetate monohydrate, Acetic acid copper (II) salt monohydrate, Copper(II) acetate hydrate

A509511-25G A509511-100G A509511-500G A509511-1000G
76695-738EA 5.31 USD
76695-738 76695-740 77572-624 77602-178 77880-104
Copper(II) acetate monohydrate 97%
Copper(II) acetate monohydrate

Precaución: for Research Use Only

Formula: (H₃CCOO)₂Cu·H₂O
MW: 199.65 g/mol
Melting Pt: 115
MDL Number: MFCD00149570
CAS Number: 6046-93-1
UN: 3077
ADR: 9 Ⅲ
Merck Index: 13,02651

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR