cis-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: cis-4-((tert-Butoxycarbonyl)amino)cyclohexanecarboxylic acid, cis-4-(BOC-amino)cyclohexanecarboxylic acid, cis-4-(Boc-amino)cyclohexanecarboxylic Acid

A504120-1G A504120-5G
76695-726EA 16 USD
76695-726 76695-728
cis-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid 97%
cis-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
MDL Number: MFCD01862294
CAS Number: 53292-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR