Carbimazole 97%

Supplier: Ambeed
Warning

Synonyms: 1-Ethoxycarbonyl-3-methyl-2-thioxo-4-imidazoline, Ethyl 3-methyl-2-thioxo-2,3-dihydro-1H-imidazole-1-carboxylate

A502733-1G A502733-250MG A502733-25G A502733-100MG
76695-718EA 18.04 USD
76695-718 76695-720 76695-722 77627-000
Carbimazole 97%
Carbimazole

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂S
MW: 186.23 g/mol
MDL Number: MFCD00027421
CAS Number: 22232-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR