4-Fluoro-3-(trifluoromethyl)benzylamine 95%

Supplier: Ambeed
Danger

Synonyms: (4-Fluoro-3-(trifluoromethyl)phenyl)methanamine

A499982-1G A499982-25G A499982-5G
76695-692EA 9 USD
76695-692 76695-694 76695-696
4-Fluoro-3-(trifluoromethyl)benzylamine 95%
4-Fluoro-3-(trifluoromethyl)benzylamine

Precaución: for Research Use Only

Formula: C₈H₇F₄N
MDL Number: MFCD00070814
CAS Number: 67515-74-6
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR