4-(n-Butoxy)benzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 4-Butoxybenzenesulphonyl chloride, 4-Butoxybenzene-1-sulphonyl chloride, 4-Butoxyphenylsulphonyl chloride

A496323-1G A496323-250MG A496323-25G A496323-5G
76695-674EA 18.04 USD
76695-674 76695-676 76695-678 76695-680
4-(n-Butoxy)benzenesulfonyl chloride 98%
4-(n-Butoxy)benzenesulfonyl chloride

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClO₃S
MW: 248.73 g/mol
MDL Number: MFCD00052344
CAS Number: 1138-56-3
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR