1-Benzothiophene-5-carbonitrile 95%

Supplier: Ambeed
Warning

A495328-100MG A495328-1G A495328-250MG
76695-668EA 31.83 USD
76695-668 76695-670 76695-672
1-Benzothiophene-5-carbonitrile 95%
1-Benzothiophene-5-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₅NS
MW: 159.208 g/mol
MDL Number: MFCD05664404
CAS Number: 2060-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR