3',5'-Dimethoxy-4'-hydroxyacetophenone 95%

Supplier: Ambeed
Warning

A492644-1G A492644-250MG A492644-25G A492644-50MG A492644-5G A492644-100G
76695-652EA 6.37 USD
76695-652 76695-654 76695-656 76695-658 76695-660 76695-662 77880-074
3',5'-Dimethoxy-4'-hydroxyacetophenone 95%
3',5'-Dimethoxy-4'-hydroxyacetophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
MDL Number: MFCD00008748
CAS Number: 2478-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR