cis-1,4-Cyclohexanediol 98%

Supplier: Ambeed
Warning

Synonyms: cis-Cyclohexane-1,4-diol, cis-1,4-Dihydroxycyclohexane

A490124-100MG A490124-1G A490124-250MG A490124-5G A490124-25G
76695-612EA 26 USD
76695-612 76695-614 76695-616 76695-618 77880-067
cis-1,4-Cyclohexanediol 98%
cis-1,4-Cyclohexanediol

Precaución: for Research Use Only

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
MDL Number: MFCD00075462
CAS Number: 931-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR