Diethyl 3,4-dihydroxyfuran-2,5-dicarboxylate 95%

Supplier: Ambeed
Warning

Synonyms: 2,5-Bis(ethoxycarbonyl)-3,4-dihydroxyfuran

A489387-1G A489387-250MG A489387-5G
76695-606EA 71 USD
76695-606 76695-608 76695-610
Diethyl 3,4-dihydroxyfuran-2,5-dicarboxylate 95%
Diethyl 3,4-dihydroxyfuran-2,5-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₇
MDL Number: MFCD00092386
CAS Number: 6270-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR