3,5-Dimethoxy-4-hydroxybenzhydrazide 95%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxy-3,5-dimethoxybenzhydrazide, 4-Hydroxy-3,5-dimethoxybenzohydrazide

A486676-1G A486676-250MG A486676-5G
76695-578EA 65.78 USD
76695-578 76695-580 76695-582
3,5-Dimethoxy-4-hydroxybenzhydrazide 95%
3,5-Dimethoxy-4-hydroxybenzhydrazide

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₄
MW: 212.21 g/mol
MDL Number: MFCD00017077
CAS Number: 1443-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR