1-(((5-(4-Nitrophenyl)furan-2-yl)methylene)amino)imidazolidine-2,4-dione 98%

Supplier: Ambeed
Warning

Synonyms: Dantrolene

A486207-100MG A486207-250MG A486207-1G
76695-574EA 24.41 USD
76695-574 76695-576 77626-968
1-(((5-(4-Nitrophenyl)furan-2-yl)methylene)amino)imidazolidine-2,4-dione 98%
1-(((5-(4-Nitrophenyl)furan-2-yl)methylene)amino)imidazolidine-2,4-dione

Precaución: for Research Use Only

Formula: C₁₄H₁₀N₄O₅
MW: 314.25 g/mol
MDL Number: MFCD00867709
CAS Number: 7261-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR