Calcium stearate 6.6-7.4% (Ca basis)

Supplier: Ambeed
Warning

Synonyms: Stearic acid calcium salt

A486184-1000G A486184-100G A486184-500G
76695-568EA 15 USD
76695-568 76695-570 76695-572
Calcium stearate 6.6-7.4% (Ca basis)
Calcium stearate

Precaución: for Research Use Only

Formula: C₃₆H₇₀CaO₄
MW: 607.03 g/mol
MDL Number: MFCD00036390
CAS Number: 1592-23-0
Merck Index: 13,01708

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR