1-(2-Aminophenyl)pyrrole 98%
Supplier: Ambeed
|
Warning
|
A483626-10G
A483626-1G
A483626-25G
A483626-5G
76695-556EA
100
USD
76695-556
76695-558
76695-560
76695-562
1-(2-Aminophenyl)pyrrole 98%
1-(2-Aminophenyl)pyrrole
Precaución: for Research Use Only
Formula:
C₁₀H₁₀N₂ |
MDL Number:
MFCD00005344 CAS Number: 6025-60-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...