7-Bromobenzofuran-2-carboxylic acid 96%

Supplier: Ambeed
Warning

A482698-100MG A482698-10G A482698-1G A482698-250MG A482698-5G
76695-542EA 29.71 USD
76695-542 76695-544 76695-546 76695-548 76695-550
7-Bromobenzofuran-2-carboxylic acid 96%
7-Bromobenzofuran-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅BrO₃
MW: 241.038 g/mol
MDL Number: MFCD10699647
CAS Number: 550998-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR